2,4-Dibromo-5-fluoro-N-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

58537-500MG 58537-1G 58537-5G
77338-632EA 504.7 USD
77338-632 77354-372 77378-492
2,4-Dibromo-5-fluoro-N-methylbenzamide ≥97%
2,4-Dibromo-5-fluoro-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₆Br₂FNO
Storage Temperature: Ambient
MDL Number: MFCD34758689
CAS Number: 2901065-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR