2,4-Dibromo-5-fluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

58536-500MG 58536-1G 58536-5G
77336-044EA 374.92 USD
77336-044 77354-370 77378-490
2,4-Dibromo-5-fluorobenzamide ≥97%
2,4-Dibromo-5-fluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₄Br₂FNO
Storage Temperature: Ambient
MDL Number: MFCD28784599
CAS Number: 221528-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR