3-Chloro-4-phenylbenzeneboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2-Chloro-[1,1'-biphenyl]-4-yl)boronic acid

58495-1G 58495-5G 58495-25G
77341-878EA 106.09 USD
77341-878 77366-550 77378-486
3-Chloro-4-phenylbenzeneboronic acid ≥95%
3-Chloro-4-phenylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BClO₂
MW: 232.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23379569
CAS Number: 179381-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR