4-Bromo-2-chloro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58494-1G 58494-5G 58494-25G
77341-744EA 103 USD
77341-744 77366-548 77378-484
4-Bromo-2-chloro-1,1'-biphenyl ≥97%
4-Bromo-2-chloro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈BrCl
MW: 267.549 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07787452
CAS Number: 58775-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR