(3-Chloro-4-phenoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

58492-500MG 58492-1G 58492-5G
77330-300EA 341.96 USD
77330-300 77354-366 77378-480
(3-Chloro-4-phenoxyphenyl)(methyl)sulfane ≥97%
(3-Chloro-4-phenoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₃H₁₁ClOS
Storage Temperature: Ambient
MDL Number: MFCD34564553
CAS Number: 2734777-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR