(4-Chloro-2,5-dimethylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

58474-1G 58474-5G 58474-25G
77341-040EA 66.95 USD
77341-040 77366-544 77378-474
(4-Chloro-2,5-dimethylphenyl)(methyl)sulfane ≥97%
(4-Chloro-2,5-dimethylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁ClS
MW: 186.702 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564552
CAS Number: 2710985-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR