1-Iodo-4-methoxy-2,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

58464-500MG 58464-1G 58464-5G 58464-25G
77330-294EA 281.19 USD
77330-294 77354-360 77366-540 77388-262
1-Iodo-4-methoxy-2,5-dimethylbenzene ≥97%
1-Iodo-4-methoxy-2,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁IO
MW: 262.088 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779016
CAS Number: 90609-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR