Ethyl 4-ethoxy-2,5-dimethylbenzoate ≥97%

Supplier: AOBChem USA

Warning

58462-500MG 58462-1G 58462-5G
77339-268EA 571.65 USD
77339-268 77354-358 77378-468
Ethyl 4-ethoxy-2,5-dimethylbenzoate ≥97%
Ethyl 4-ethoxy-2,5-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31773131
CAS Number: 1368217-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR