4-Ethoxy-2,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

58460-500MG 58460-1G 58460-5G
77340-318EA 900.22 USD
77340-318 77354-354 77378-464
4-Ethoxy-2,5-dimethylbenzoic acid ≥97%
4-Ethoxy-2,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14537714
CAS Number: 482355-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR