4-Ethoxy-2,5-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58458-500MG 58458-1G 58458-5G
77338-082EA 462.47 USD
77338-082 77354-352 77378-462
4-Ethoxy-2,5-dimethylbenzaldehyde ≥97%
4-Ethoxy-2,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD06247331
CAS Number: 147351-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR