2-Fluoro-3-methoxy-5-(trifluoromethyl)-1,1'-biphenyl ≥97%
Supplier: AOBChem USA
|
Warning
|
58445-500MG
58445-1G
58445-5G
58445-25G
77330-286EA
225.57
USD
77330-286
77354-350
77366-532
77388-256
2-Fluoro-3-methoxy-5-(trifluoromethyl)-1,1'-biphenyl ≥97%
2-Fluoro-3-methoxy-5-(trifluoromethyl)-1,1'-biphenyl
Precaución: for Research Use Only
Formula:
C₁₄H₁₀F₄O MW: 270.222 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34758682 CAS Number: 2755723-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...