Methyl 5-fluoro-4-methoxy-2-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

58441-500MG 58441-1G 58441-5G
77339-966EA 694.22 USD
77339-966 77354-346 77378-456
Methyl 5-fluoro-4-methoxy-2-(trifluoromethyl)benzoate ≥97%
Methyl 5-fluoro-4-methoxy-2-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₃
MW: 252.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758678
CAS Number: 2704722-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR