5-Fluoro-4-methoxy-2-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

58440-500MG 58440-1G 58440-5G
77338-264EA 470.71 USD
77338-264 77354-344 77378-454
5-Fluoro-4-methoxy-2-(trifluoromethyl)benzoic acid ≥97%
5-Fluoro-4-methoxy-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₆F₄O₃
MW: 238.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758677
CAS Number: 2090520-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR