(5-Fluoro-4-methoxy-2-(trifluoromethyl)phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

58439-500MG 58439-1G 58439-5G
77339-964EA 694.22 USD
77339-964 77354-342 77378-452
(5-Fluoro-4-methoxy-2-(trifluoromethyl)phenyl)methanol ≥95%
(5-Fluoro-4-methoxy-2-(trifluoromethyl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₈F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD34758676
CAS Number: 2091608-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR