1-Bromo-4-ethoxy-5-fluoro-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

58435-500MG 58435-1G 58435-5G
77336-638EA 390.37 USD
77336-638 77354-338 77378-448
1-Bromo-4-ethoxy-5-fluoro-2-(trifluoromethyl)benzene ≥97%
1-Bromo-4-ethoxy-5-fluoro-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₇BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD34758672
CAS Number: 2385897-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR