1-(Benzyloxy)-4-bromo-2-fluoro-5-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

58434-500MG 58434-1G 58434-5G
77340-138EA 791.04 USD
77340-138 77354-336 77378-446
1-(Benzyloxy)-4-bromo-2-fluoro-5-(trifluoromethyl)benzene ≥95%
1-(Benzyloxy)-4-bromo-2-fluoro-5-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₉BrF₄O
MW: 349.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758671
CAS Number: 2807455-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR