1-Bromo-5-fluoro-4-isopropoxy-2-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

58433-500MG 58433-1G 58433-5G 58433-25G
77330-284EA 172.01 USD
77330-284 77346-418 77366-530 77388-254
1-Bromo-5-fluoro-4-isopropoxy-2-(trifluoromethyl)benzene ≥97%
1-Bromo-5-fluoro-4-isopropoxy-2-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₉BrF₄O
MW: 301.075 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758670
CAS Number: 2807466-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR