Ethyl 2-fluoro-3-hydroxy-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

58429-500MG 58429-1G 58429-5G
77330-282EA 328.57 USD
77330-282 77354-334 77378-444
Ethyl 2-fluoro-3-hydroxy-5-(trifluoromethyl)benzoate ≥97%
Ethyl 2-fluoro-3-hydroxy-5-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₃
MW: 252.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758666
CAS Number: 2701608-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR