2-Fluoro-3-hydroxy-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

58427-500MG 58427-1G 58427-5G 58427-25G
77330-280EA 224.54 USD
77330-280 77354-332 77366-528 77388-252
2-Fluoro-3-hydroxy-5-(trifluoromethyl)benzoic acid ≥97%
2-Fluoro-3-hydroxy-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34758664
CAS Number: 2385182-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR