2-Fluoro-3-(hydroxymethyl)-5-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

58425-500MG 58425-1G 58425-5G
77335-428EA 353.29 USD
77335-428 77354-330 77378-442
2-Fluoro-3-(hydroxymethyl)-5-(trifluoromethyl)phenol ≥97%
2-Fluoro-3-(hydroxymethyl)-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₈H₆F₄O₂
MW: 210.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758662
CAS Number: 2764728-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR