2-Fluoro-3-iodo-5-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

58421-500MG 58421-1G 58421-5G
77330-276EA 342.99 USD
77330-276 77354-326 77378-440
2-Fluoro-3-iodo-5-(trifluoromethyl)phenol ≥97%
2-Fluoro-3-iodo-5-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₃F₄IO
MW: 305.996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758658
CAS Number: 2386337-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR