2-Isopropyl-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58392-500MG 58392-1G 58392-5G
77337-456EA 414.06 USD
77337-456 77354-324 77378-438
2-Isopropyl-4-(methylthio)benzaldehyde ≥97%
2-Isopropyl-4-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₄OS
MW: 194.293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29121128
CAS Number: 1289090-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR