2-Chloro-3-methoxy-4-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

58391-500MG 58391-1G 58391-25G 58391-5G
77330-274EA 292.52 USD
77330-274 77354-322 77388-248 77378-436
2-Chloro-3-methoxy-4-phenylpyridine ≥97%
2-Chloro-3-methoxy-4-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD28753528
CAS Number: 1805229-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR