4-Chloro-4'-fluoro-3,5-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58379-500MG 58379-1G 58379-5G 58379-25G
77330-270EA 198.79 USD
77330-270 77354-316 77366-522 77388-244
4-Chloro-4'-fluoro-3,5-dimethyl-1,1'-biphenyl ≥97%
4-Chloro-4'-fluoro-3,5-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClF
MW: 234.696 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758622
CAS Number: 2755721-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR