4-Chloro-3,4',5-trimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58378-500MG 58378-1G 58378-5G 58378-25G
77330-268EA 229.69 USD
77330-268 77354-314 77366-520 77388-242
4-Chloro-3,4',5-trimethyl-1,1'-biphenyl ≥97%
4-Chloro-3,4',5-trimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₅Cl
MW: 230.733 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758621
CAS Number: 2408229-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR