4'-Methoxy-3',5'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
58352-500MG
58352-1G
58352-5G
58352-25G
77330-266EA
145.23
USD
77330-266
77345-094
77366-518
77388-240
4'-Methoxy-3',5'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
4'-Methoxy-3',5'-dimethyl-[1,1'-biphenyl]-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₆H₁₆O₂ MW: 240.297 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD25866125 CAS Number: 154657-70-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...