4-Methoxy-3,5-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58348-500MG 58348-1G 58348-5G 58348-25G
77330-260EA 126.69 USD
77330-260 77344-154 77366-512 77378-422
4-Methoxy-3,5-dimethyl-1,1'-biphenyl ≥97%
4-Methoxy-3,5-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₆O
MW: 212.287 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758595
CAS Number: 94001-39-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR