3-Ethoxy-5-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

58327-500MG 58327-1G 58327-5G
77330-258EA 340.93 USD
77330-258 77354-300 77378-414
3-Ethoxy-5-formylbenzoic acid ≥97%
3-Ethoxy-5-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD34758584
CAS Number: 2807448-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR