3-Ethoxy-5-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

58325-500MG 58325-1G 58325-5G 58325-25G
77330-256EA 204.97 USD
77330-256 77354-298 77366-510 77388-236
3-Ethoxy-5-(methylthio)benzaldehyde ≥97%
3-Ethoxy-5-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂S
MW: 196.266 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758582
CAS Number: 2901102-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR