5'-Hydroxy-2'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

58322-500MG 58322-1G 58322-5G 58322-25G
77330-254EA 281.19 USD
77330-254 77354-296 77366-508 77388-234
5'-Hydroxy-2'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
5'-Hydroxy-2'-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MW: 212.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758579
CAS Number: 2764730-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR