4'-Fluoro-6-methyl-[1,1'-biphenyl]-3-ol ≥97%

Supplier: AOBChem USA

Warning

58321-500MG 58321-1G 58321-5G
77335-426EA 353.29 USD
77335-426 77354-294 77378-412
4'-Fluoro-6-methyl-[1,1'-biphenyl]-3-ol ≥97%
4'-Fluoro-6-methyl-[1,1'-biphenyl]-3-ol

Precaución: for Research Use Only

Formula: C₁₃H₁₁FO
MW: 202.224 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758578
CAS Number: 1856797-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR