4',6-Dimethyl-[1,1'-biphenyl]-3-ol ≥97%

Supplier: AOBChem USA

Warning

58320-500MG 58320-1G 58320-5G
77330-252EA 334.75 USD
77330-252 77354-292 77378-410
4',6-Dimethyl-[1,1'-biphenyl]-3-ol ≥97%
4',6-Dimethyl-[1,1'-biphenyl]-3-ol

Precaución: for Research Use Only

Formula: C₁₄H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD34758577
CAS Number: 1861551-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR