4'-Fluoro-2-methyl-[1,1'-biphenyl]-3-amine ≥97%

Supplier: AOBChem USA

Warning

58317-500MG 58317-1G 58317-5G 58317-25G
77330-250EA 231.75 USD
77330-250 77354-290 77366-506 77388-232
4'-Fluoro-2-methyl-[1,1'-biphenyl]-3-amine ≥97%
4'-Fluoro-2-methyl-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₂FN
MW: 201.239 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31863148
CAS Number: 1694468-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR