2,4'-Dimethyl-[1,1'-biphenyl]-3-amine ≥97%

Supplier: AOBChem USA

Warning

58316-500MG 58316-1G 58316-5G
77330-248EA 315.18 USD
77330-248 77354-288 77378-408
2,4'-Dimethyl-[1,1'-biphenyl]-3-amine ≥97%
2,4'-Dimethyl-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₅N
MW: 197.276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14585569
CAS Number: 637353-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR