3-Bromo-4'-fluoro-2-methyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

58313-500MG 58313-1G 58313-5G 58313-25G
77330-246EA 225.57 USD
77330-246 77354-286 77366-504 77388-230
3-Bromo-4'-fluoro-2-methyl-1,1'-biphenyl ≥95%
3-Bromo-4'-fluoro-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrF
MW: 265.121 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758576
CAS Number: 2764732-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR