3-Bromo-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58310-500MG 58310-1G 58310-5G 58310-25G
77330-242EA 234.84 USD
77330-242 77354-282 77366-500 77388-226
3-Bromo-2-methyl-1,1'-biphenyl ≥97%
3-Bromo-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁Br
Storage Temperature: Ambient
MDL Number: MFCD29923329
CAS Number: 172976-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR