3'-Chloro-2'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

58309-500MG 58309-1G 58309-5G 58309-25G
77330-240EA 275.01 USD
77330-240 77354-280 77366-498 77388-224
3'-Chloro-2'-methyl-[1,1'-biphenyl]-4-carbaldehyde ≥97%
3'-Chloro-2'-methyl-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁ClO
MW: 230.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11895761
CAS Number: 1181295-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR