3-Chloro-4'-fluoro-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58308-500MG 58308-1G 58308-5G
77337-520EA 416.12 USD
77337-520 77354-278 77378-406
3-Chloro-4'-fluoro-2-methyl-1,1'-biphenyl ≥97%
3-Chloro-4'-fluoro-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀ClF
MW: 220.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14701443
CAS Number: 1214327-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR