3-Chloro-2,4'-dimethyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58306-500MG 58306-1G 58306-5G
77337-454EA 414.06 USD
77337-454 77354-276 77378-404
3-Chloro-2,4'-dimethyl-1,1'-biphenyl ≥97%
3-Chloro-2,4'-dimethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃Cl
Storage Temperature: Ambient
MDL Number: MFCD34758574
CAS Number: 2755719-76-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR