3-Chloro-2-methyl-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58305-500MG 58305-1G 58305-5G 58305-25G
77330-238EA 198.79 USD
77330-238 77354-274 77366-496 77388-222
3-Chloro-2-methyl-1,1'-biphenyl ≥97%
3-Chloro-2-methyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁Cl
Storage Temperature: Ambient
MDL Number: MFCD00134199
CAS Number: 20261-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR