1,3-Diiodo-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

58304-500MG 58304-1G 58304-5G 58304-25G
77330-236EA 103 USD
77330-236 77343-034 77366-494 77378-402
1,3-Diiodo-2-methylbenzene ≥97%
1,3-Diiodo-2-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₆I₂
Storage Temperature: Ambient
MDL Number: MFCD22571073
CAS Number: 89795-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR