2'-Methyl-3'-nitro-[1,1'-biphenyl]-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

58303-500MG 58303-1G 58303-5G 58303-25G
77330-234EA 225.57 USD
77330-234 77354-272 77366-492 77388-220
2'-Methyl-3'-nitro-[1,1'-biphenyl]-4-carbaldehyde ≥97%
2'-Methyl-3'-nitro-[1,1'-biphenyl]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₁NO₃
MW: 241.242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758573
CAS Number: 2764728-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR