4'-Fluoro-2-methyl-3-nitro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58302-500MG 58302-1G 58302-5G 58302-25G
77330-232EA 262.65 USD
77330-232 77354-270 77366-490 77388-218
4'-Fluoro-2-methyl-3-nitro-1,1'-biphenyl ≥97%
4'-Fluoro-2-methyl-3-nitro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀FNO₂
MW: 231.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758572
CAS Number: 2755720-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR