2,4'-Dimethyl-3-nitro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58301-500MG 58301-1G 58301-5G 58301-25G
77330-230EA 269.86 USD
77330-230 77354-268 77366-488 77388-216
2,4'-Dimethyl-3-nitro-1,1'-biphenyl ≥97%
2,4'-Dimethyl-3-nitro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂
MW: 227.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758571
CAS Number: 2755722-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR