4',6-Dimethyl-[1,1'-biphenyl]-3-amine ≥97%

Supplier: AOBChem USA

Warning

58296-500MG 58296-1G 58296-5G
77330-224EA 319.3 USD
77330-224 77354-262 77378-398
4',6-Dimethyl-[1,1'-biphenyl]-3-amine ≥97%
4',6-Dimethyl-[1,1'-biphenyl]-3-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₅N
MW: 197.276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14585559
CAS Number: 1208082-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR