4'-Fluoro-2-methyl-5-nitro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58295-500MG 58295-1G 58295-5G 58295-25G
77330-222EA 187.46 USD
77330-222 77354-260 77366-484 77388-212
4'-Fluoro-2-methyl-5-nitro-1,1'-biphenyl ≥97%
4'-Fluoro-2-methyl-5-nitro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₀FNO₂
MW: 231.222 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18207476
CAS Number: 223767-01-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR