2,4'-Dimethyl-5-nitro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

58294-500MG 58294-1G 58294-5G 58294-25G
77330-220EA 187.46 USD
77330-220 77354-258 77366-482 77388-210
2,4'-Dimethyl-5-nitro-1,1'-biphenyl ≥97%
2,4'-Dimethyl-5-nitro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂
MW: 227.259 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18207478
CAS Number: 70689-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR