5-(2-Methoxy-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58287-500MG 58287-1G 58287-5G
77337-248EA 409.94 USD
77337-248 77354-256 77378-396
5-(2-Methoxy-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(2-Methoxy-5-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₉F₃N₂OS
MW: 274.262 g/mol
Storage Temperature: Ambient
CAS Number: 2901103-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR