(3-Bromo-5-(trifluoromethoxy)phenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

58269-500MG 58269-1G 58269-5G 58269-25G
77330-216EA 231.75 USD
77330-216 77354-252 77366-480 77388-206
(3-Bromo-5-(trifluoromethoxy)phenyl)(thiomorpholino)methanone ≥97%
(3-Bromo-5-(trifluoromethoxy)phenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₁BrF₃NO₂S
MW: 370.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758562
CAS Number: 2807469-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR