(3-Bromo-5-(trifluoromethoxy)phenyl)(morpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

58268-500MG 58268-1G 58268-5G 58268-25G
77330-214EA 231.75 USD
77330-214 77354-250 77366-478 77388-204
(3-Bromo-5-(trifluoromethoxy)phenyl)(morpholino)methanone ≥97%
(3-Bromo-5-(trifluoromethoxy)phenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₁BrF₃NO₃
MW: 354.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34758561
CAS Number: 2807454-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR